Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 5-phenoxycarbonylamino-1,2,3-thiadiazole-4-carboxylate, 98+%

$295.54 - $295.54

Chemical Identifiers

Chemical Name or Material Ethyl 5-phenoxycarbonylamino-1,2,3-thiadiazole-4-carboxylate
CAS 2037-81-2
Molecular Formula C12H11N3O4S
Formula Weight 293.3
MDL Number MFCD00100230
Synonym 5-Amino-1,2,3-thiadiazole-4-carboxylic acid ethyl ester 5-(phenyl carbamate; 5-Phenoxycarbonylamino-1,2,3-thiadiazole-4-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH25763MD

 
alfa aesar™
H25763MD
250mg Each for $295.54
Description & Specifications

Specifications

Formula Weight 293.3
Chemical Name or Material Ethyl 5-phenoxycarbonylamino-1,2,3-thiadiazole-4-carboxylate
Assay ≥98%
Molecular Formula C12H11N3O4S
MDL Number MFCD00100230
CAS 2037-81-2
Synonym 5-Amino-1,2,3-thiadiazole-4-carboxylic acid ethyl ester 5-(phenyl carbamate; 5-Phenoxycarbonylamino-1,2,3-thiadiazole-4-carboxylic acid ethyl ester
Melting Point 153° to 155°C
Quantity 250mg