Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 5-methylisoxazole-4-carboxylate, 96%

$44.31 - $142.53

Chemical Identifiers

Chemical Name or Material Ethyl 5-methylisoxazole-4-carboxylate
CAS 51135-73-0
Molecular Formula C7H9NO3
Formula Weight 155.15
MDL Number MFCD01631119
Beilstein 122097
Synonym 5-Methylisoxazole-4-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1864704

  SDS
alfa aesar™
L1864704
2g Each for $44.31

AAL1864709

  SDS
alfa aesar™
L1864709
10g Each for $142.53
Description & Specifications

Specifications

Formula Weight 155.15
Synonym 5-Methylisoxazole-4-carboxylic acid ethyl ester
CAS 51135-73-0
Molecular Formula C7H9NO3
Beilstein 122097
MDL Number MFCD01631119
Health Hazard 1 Warning
Assay 96%
Boiling Point 95° to 97°C (15mmHg)
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Quantity 2g
Chemical Name or Material Ethyl 5-methylisoxazole-4-carboxylate
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.