Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 5-methylisoxazole-3-carboxylate, 95%

$50.78 - $192.86

Chemical Identifiers

Chemical Name or Material Ethyl 5-methylisoxazole-3-carboxylate
CAS 3209-72-1
Molecular Formula C7H9NO3
Formula Weight 155.15
MDL Number MFCD00085080
Synonym 5-Methylisoxazole-3-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1322603

  SDS
alfa aesar™
A1322603
1g Each for $50.78

AAA1322606

  SDS
alfa aesar™
A1322606
5g Each for $192.86

AAA1322614

  SDS
alfa aesar™
A1322614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Ethyl 5-methylisoxazole-3-carboxylate
CAS 3209-72-1
Assay 95%
Molecular Formula C7H9NO3
Formula Weight 155.15
MDL Number MFCD00085080
Synonym 5-Methylisoxazole-3-carboxylic acid ethyl ester
Boiling Point 77° to 79°C (0.5mmHg)
Melting Point 27° to 31°C
Quantity 1g
Flash Point 109°C (228°F)

Applications
Ethyl 5-methylisoxazole-3-carboxylate, It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents.