missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 5-methyl-4-oxo-3,4-dihydropyrrolo[2,1-f]-1,2,4-triazine-6-carboxylate, 97%, Alfa Aesar™

$190.43 - $571.30

Chemical Identifiers

CAS 427878-70-4
Molecular Formula C10H11N3O3
Molecular Weight (g/mol) 221.216
MDL Number MFCD11044884
InChI Key XCUHVGKOOKXZNK-UHFFFAOYSA-N
Synonym 5-methyl-4-oxo-3,4-dihydro-pyrrolo 2,1-f 1,2,4 triazine-6-carboxylic acid ethyl ester, ethyl 4-hydroxy-5-methylpyrrolo 1,2-f 1,2,4 triazine-6-carboxylate, ethyl 4-hydroxy-5-methylpyrrolo 2,1-f 1,2,4 triazine-6-carboxylate, ethyl 5-methyl-4-oxo-1,4-dihydropyrrolo 2,1-f 1,2,4 triazine-6-carboxylate, ethyl 5-methyl-4-oxo-1h-pyrrolo 2,1-f 1,2,4 triazine-6-carboxylate, ethyl 5-methyl-4-oxo-3,4-dihydropyrrolo 1,2-f 1,2,4 triazine-6-carboxylate, ethyl 5-methyl-4-oxo-3,4-dihydropyrrolo 2,1-f 1,2,4 triazine-6-carboxylate, ethyl 5-methyl-4-oxo-3h,4h-pyrrolo 2,1-f 1,2,4 triazine-6-carboxylate, ethyl 5-methyl-4-oxo-3h-pyrrolo 2,1-f 1,2,4 triazine-6-carboxylate, pyrrolo 2,1-f 1,2,4 triazine-6-carboxylic acid, 1,4-dihydro-5-methyl-4-oxo-, ethyl ester
PubChem CID 11572090
IUPAC Name ethyl 5-methyl-4-oxo-1H-pyrrolo[2,1-f][1,2,4]triazine-6-carboxylate
SMILES CCOC(=O)C1=CN2C(=C1C)C(=O)N=CN2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH65101MD
View Documents
Thermo Fisher Scientific
H65101MD
250mg Each for $190.43
 
AAH6510103
View Documents
Thermo Fisher Scientific
H6510103
1g Each for $571.30
 
Specifications

Chemical Identifiers

427878-70-4
221.216
XCUHVGKOOKXZNK-UHFFFAOYSA-N
11572090
CCOC(=O)C1=CN2C(=C1C)C(=O)N=CN2
C10H11N3O3
MFCD11044884
5-methyl-4-oxo-3,4-dihydro-pyrrolo 2,1-f 1,2,4 triazine-6-carboxylic acid ethyl ester, ethyl 4-hydroxy-5-methylpyrrolo 1,2-f 1,2,4 triazine-6-carboxylate, ethyl 4-hydroxy-5-methylpyrrolo 2,1-f 1,2,4 triazine-6-carboxylate, ethyl 5-methyl-4-oxo-1,4-dihydropyrrolo 2,1-f 1,2,4 triazine-6-carboxylate, ethyl 5-methyl-4-oxo-1h-pyrrolo 2,1-f 1,2,4 triazine-6-carboxylate, ethyl 5-methyl-4-oxo-3,4-dihydropyrrolo 1,2-f 1,2,4 triazine-6-carboxylate, ethyl 5-methyl-4-oxo-3,4-dihydropyrrolo 2,1-f 1,2,4 triazine-6-carboxylate, ethyl 5-methyl-4-oxo-3h,4h-pyrrolo 2,1-f 1,2,4 triazine-6-carboxylate, ethyl 5-methyl-4-oxo-3h-pyrrolo 2,1-f 1,2,4 triazine-6-carboxylate, pyrrolo 2,1-f 1,2,4 triazine-6-carboxylic acid, 1,4-dihydro-5-methyl-4-oxo-, ethyl ester
ethyl 5-methyl-4-oxo-1H-pyrrolo[2,1-f][1,2,4]triazine-6-carboxylate

Specifications

Yellow
427878-70-4
MFCD11044884
5-methyl-4-oxo-3,4-dihydro-pyrrolo 2,1-f 1,2,4 triazine-6-carboxylic acid ethyl ester, ethyl 4-hydroxy-5-methylpyrrolo 1,2-f 1,2,4 triazine-6-carboxylate, ethyl 4-hydroxy-5-methylpyrrolo 2,1-f 1,2,4 triazine-6-carboxylate, ethyl 5-methyl-4-oxo-1,4-dihydropyrrolo 2,1-f 1,2,4 triazine-6-carboxylate, ethyl 5-methyl-4-oxo-1h-pyrrolo 2,1-f 1,2,4 triazine-6-carboxylate, ethyl 5-methyl-4-oxo-3,4-dihydropyrrolo 1,2-f 1,2,4 triazine-6-carboxylate, ethyl 5-methyl-4-oxo-3,4-dihydropyrrolo 2,1-f 1,2,4 triazine-6-carboxylate, ethyl 5-methyl-4-oxo-3h,4h-pyrrolo 2,1-f 1,2,4 triazine-6-carboxylate, ethyl 5-methyl-4-oxo-3h-pyrrolo 2,1-f 1,2,4 triazine-6-carboxylate, pyrrolo 2,1-f 1,2,4 triazine-6-carboxylic acid, 1,4-dihydro-5-methyl-4-oxo-, ethyl ester
CCOC(=O)C1=CN2C(=C1C)C(=O)N=CN2
221.216
221.22
97%
250mg
C10H11N3O3
10162668
XCUHVGKOOKXZNK-UHFFFAOYSA-N
ethyl 5-methyl-4-oxo-1H-pyrrolo[2,1-f][1,2,4]triazine-6-carboxylate
11572090
Powder
Ethyl 5-methyl-4-oxo-3,4-dihydropyrrolo[2,1-f]-1,2,4-triazine-6-carboxylate
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures