missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 5-formyl-2,4-dimethylpyrrole-3-carboxylate, 95%

$102.41 - $306.88

Chemical Identifiers

CAS 2199-59-9
Molecular Formula C10H13NO3
Molecular Weight (g/mol) 195.218
MDL Number MFCD00030352
InChI Key GDISALBEIGGPER-UHFFFAOYSA-N
Synonym 5-formyl-2,4-dimethyl-1h-pyrrole-3-carboxylic acid ethyl ester, 2,4-dimethyl-3-ethoxycarbonyl-5-formylpyrrole, ethyl 2,4-dimethyl-5-formylpyrrole-3-carboxylate, ethyl 5-formyl-2,4-dimethylpyrrole-3-carboxylate, 1h-pyrrole-3-carboxylic acid, 5-formyl-2,4-dimethyl-, ethyl ester, ethyl 3,5-dimethyl-2-formylpyrrole-4-carboxylate, 2,4-dimethyl-3-carbethoxy pyrrole-5-carboxaldehyde, 3,5-dimethyl-4-ethoxycarbonyl-1h-pyrrole-2-carboxaldehyde, 5-formyl-2,4-dimethyl-3-pyrrolecarboxylic acid ethyl ester, pubchem9415
PubChem CID 137485
IUPAC Name ethyl 5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxylate
SMILES CCOC(=O)C1=C(NC(=C1C)C=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63751MD View Documents Alfa Aesar™
H63751MD
250mg Each for $102.41
 
AAH6375103 View Documents Alfa Aesar™
H6375103
1g Each for $306.88
 
Specifications

Chemical Identifiers

2199-59-9
195.218
GDISALBEIGGPER-UHFFFAOYSA-N
137485
CCOC(=O)C1=C(NC(=C1C)C=O)C
C10H13NO3
MFCD00030352
5-formyl-2,4-dimethyl-1h-pyrrole-3-carboxylic acid ethyl ester, 2,4-dimethyl-3-ethoxycarbonyl-5-formylpyrrole, ethyl 2,4-dimethyl-5-formylpyrrole-3-carboxylate, ethyl 5-formyl-2,4-dimethylpyrrole-3-carboxylate, 1h-pyrrole-3-carboxylic acid, 5-formyl-2,4-dimethyl-, ethyl ester, ethyl 3,5-dimethyl-2-formylpyrrole-4-carboxylate, 2,4-dimethyl-3-carbethoxy pyrrole-5-carboxaldehyde, 3,5-dimethyl-4-ethoxycarbonyl-1h-pyrrole-2-carboxaldehyde, 5-formyl-2,4-dimethyl-3-pyrrolecarboxylic acid ethyl ester, pubchem9415
ethyl 5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxylate

Specifications

250mg
C10H13NO3
5-formyl-2,4-dimethyl-1h-pyrrole-3-carboxylic acid ethyl ester, 2,4-dimethyl-3-ethoxycarbonyl-5-formylpyrrole, ethyl 2,4-dimethyl-5-formylpyrrole-3-carboxylate, ethyl 5-formyl-2,4-dimethylpyrrole-3-carboxylate, 1h-pyrrole-3-carboxylic acid, 5-formyl-2,4-dimethyl-, ethyl ester, ethyl 3,5-dimethyl-2-formylpyrrole-4-carboxylate, 2,4-dimethyl-3-carbethoxy pyrrole-5-carboxaldehyde, 3,5-dimethyl-4-ethoxycarbonyl-1h-pyrrole-2-carboxaldehyde, 5-formyl-2,4-dimethyl-3-pyrrolecarboxylic acid ethyl ester, pubchem9415
CCOC(=O)C1=C(NC(=C1C)C=O)C
195.218
195.22
95%
2199-59-9
MFCD00030352
GDISALBEIGGPER-UHFFFAOYSA-N
ethyl 5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxylate
137485
Solid
Ethyl 5-formyl-2,4-dimethylpyrrole-3-carboxylate