missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 5-formyl-2,4-dimethylpyrrole-3-carboxylate, 95%

Manufacturer:  Alfa Aesar™ H63751MD

 View more versions of this product

Catalog No. AAH63751MD



Specifications

Specifications

Ethyl 5-formyl-2,4-dimethylpyrrole-3-carboxylate
2199-59-9
MFCD00030352
GDISALBEIGGPER-UHFFFAOYSA-N
ethyl 5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxylate
137485
Solid
250mg
C10H13NO3
5-formyl-2,4-dimethyl-1h-pyrrole-3-carboxylic acid ethyl ester, 2,4-dimethyl-3-ethoxycarbonyl-5-formylpyrrole, ethyl 2,4-dimethyl-5-formylpyrrole-3-carboxylate, ethyl 5-formyl-2,4-dimethylpyrrole-3-carboxylate, 1h-pyrrole-3-carboxylic acid, 5-formyl-2,4-dimethyl-, ethyl ester, ethyl 3,5-dimethyl-2-formylpyrrole-4-carboxylate, 2,4-dimethyl-3-carbethoxy pyrrole-5-carboxaldehyde, 3,5-dimethyl-4-ethoxycarbonyl-1h-pyrrole-2-carboxaldehyde, 5-formyl-2,4-dimethyl-3-pyrrolecarboxylic acid ethyl ester, pubchem9415
CCOC(=O)C1=C(NC(=C1C)C=O)C
195.218
195.22
95%

Chemical Identifiers

2199-59-9
195.218
GDISALBEIGGPER-UHFFFAOYSA-N
137485
CCOC(=O)C1=C(NC(=C1C)C=O)C
C10H13NO3
MFCD00030352
5-formyl-2,4-dimethyl-1h-pyrrole-3-carboxylic acid ethyl ester, 2,4-dimethyl-3-ethoxycarbonyl-5-formylpyrrole, ethyl 2,4-dimethyl-5-formylpyrrole-3-carboxylate, ethyl 5-formyl-2,4-dimethylpyrrole-3-carboxylate, 1h-pyrrole-3-carboxylic acid, 5-formyl-2,4-dimethyl-, ethyl ester, ethyl 3,5-dimethyl-2-formylpyrrole-4-carboxylate, 2,4-dimethyl-3-carbethoxy pyrrole-5-carboxaldehyde, 3,5-dimethyl-4-ethoxycarbonyl-1h-pyrrole-2-carboxaldehyde, 5-formyl-2,4-dimethyl-3-pyrrolecarboxylic acid ethyl ester, pubchem9415
ethyl 5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxylate
SDS Documents