missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl 5-Formyl-2,4-dimethyl-3-pyrrolecarboxylate 98.0+%, TCI America™
$86.27 - $340.10
Chemical Identifiers
CAS | 2199-59-9 |
---|---|
Molecular Formula | C10H13NO3 |
Molecular Weight (g/mol) | 195.22 |
MDL Number | MFCD00030352 |
InChI Key | GDISALBEIGGPER-UHFFFAOYSA-N |
Synonym | 5-formyl-2,4-dimethyl-1h-pyrrole-3-carboxylic acid ethyl ester, 2,4-dimethyl-3-ethoxycarbonyl-5-formylpyrrole, ethyl 2,4-dimethyl-5-formylpyrrole-3-carboxylate, ethyl 5-formyl-2,4-dimethylpyrrole-3-carboxylate, 1h-pyrrole-3-carboxylic acid, 5-formyl-2,4-dimethyl-, ethyl ester, ethyl 3,5-dimethyl-2-formylpyrrole-4-carboxylate, 2,4-dimethyl-3-carbethoxy pyrrole-5-carboxaldehyde, 3,5-dimethyl-4-ethoxycarbonyl-1h-pyrrole-2-carboxaldehyde, 5-formyl-2,4-dimethyl-3-pyrrolecarboxylic acid ethyl ester, pubchem9415 |
PubChem CID | 137485 |
IUPAC Name | ethyl 5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxylate |
SMILES | CCOC(=O)C1=C(C)NC(C=O)=C1C |
Chemical Identifiers
2199-59-9 | |
195.22 | |
GDISALBEIGGPER-UHFFFAOYSA-N | |
137485 | |
CCOC(=O)C1=C(C)NC(C=O)=C1C |
C10H13NO3 | |
MFCD00030352 | |
5-formyl-2,4-dimethyl-1h-pyrrole-3-carboxylic acid ethyl ester, 2,4-dimethyl-3-ethoxycarbonyl-5-formylpyrrole, ethyl 2,4-dimethyl-5-formylpyrrole-3-carboxylate, ethyl 5-formyl-2,4-dimethylpyrrole-3-carboxylate, 1h-pyrrole-3-carboxylic acid, 5-formyl-2,4-dimethyl-, ethyl ester, ethyl 3,5-dimethyl-2-formylpyrrole-4-carboxylate, 2,4-dimethyl-3-carbethoxy pyrrole-5-carboxaldehyde, 3,5-dimethyl-4-ethoxycarbonyl-1h-pyrrole-2-carboxaldehyde, 5-formyl-2,4-dimethyl-3-pyrrolecarboxylic acid ethyl ester, pubchem9415 | |
ethyl 5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxylate |
Specifications
2199-59-9 | |
White-Yellow | |
MFCD00030352 | |
5-formyl-2,4-dimethyl-1h-pyrrole-3-carboxylic acid ethyl ester, 2,4-dimethyl-3-ethoxycarbonyl-5-formylpyrrole, ethyl 2,4-dimethyl-5-formylpyrrole-3-carboxylate, ethyl 5-formyl-2,4-dimethylpyrrole-3-carboxylate, 1h-pyrrole-3-carboxylic acid, 5-formyl-2,4-dimethyl-, ethyl ester, ethyl 3,5-dimethyl-2-formylpyrrole-4-carboxylate, 2,4-dimethyl-3-carbethoxy pyrrole-5-carboxaldehyde, 3,5-dimethyl-4-ethoxycarbonyl-1h-pyrrole-2-carboxaldehyde, 5-formyl-2,4-dimethyl-3-pyrrolecarboxylic acid ethyl ester, pubchem9415 | |
CCOC(=O)C1=C(C)NC(C=O)=C1C | |
195.22 | |
195.22 | |
Crystalline Powder |
166°C | |
C10H13NO3 | |
1 g | |
GDISALBEIGGPER-UHFFFAOYSA-N | |
ethyl 5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxylate | |
137485 | |
≥98.0% (GC) | |
Ethyl 5-Formyl-2,4-dimethyl-3-pyrrolecarboxylate |
Safety and Handling
TSCA : No