missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl 5-chlorothiophene-2-carboxylate, 98+%, Thermo Scientific™
$64.26 - $64.26
Chemical Identifiers
CAS | 5751-82-6 |
---|---|
Molecular Formula | C7H7ClO2S |
Molecular Weight (g/mol) | 190.641 |
MDL Number | MFCD00051732 |
InChI Key | BMOKMXXTOZFEIZ-UHFFFAOYSA-N |
Synonym | 5-chloro-2-thiophenecarboxylic acid ethyl ester, 5-chlorothiophene-2-carboxylic acid ethyl ester, 2-thiophenecarboxylic acid, 5-chloro-, ethyl ester, ethyl 5-chlorothiophene-2-carboxate, ethyl 5-chloro-2-thiophenecarboxylate, pubchem16133, acmc-209znt, ksc494s8t, ethyl5-chlorothiophene-2-carboxlate, ethyl 5-chloranylthiophene-2-carboxylate |
PubChem CID | 521808 |
IUPAC Name | ethyl 5-chlorothiophene-2-carboxylate |
SMILES | CCOC(=O)C1=CC=C(S1)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0638503
|
Thermo Scientific Chemicals
L0638503 |
1 g |
Each for $64.26
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
5751-82-6 | |
190.641 | |
BMOKMXXTOZFEIZ-UHFFFAOYSA-N | |
521808 | |
CCOC(=O)C1=CC=C(S1)Cl |
C7H7ClO2S | |
MFCD00051732 | |
5-chloro-2-thiophenecarboxylic acid ethyl ester, 5-chlorothiophene-2-carboxylic acid ethyl ester, 2-thiophenecarboxylic acid, 5-chloro-, ethyl ester, ethyl 5-chlorothiophene-2-carboxate, ethyl 5-chloro-2-thiophenecarboxylate, pubchem16133, acmc-209znt, ksc494s8t, ethyl5-chlorothiophene-2-carboxlate, ethyl 5-chloranylthiophene-2-carboxylate | |
ethyl 5-chlorothiophene-2-carboxylate |
Specifications
5751-82-6 | |
C7H7ClO2S | |
MFCD00051732 | |
128707 | |
BMOKMXXTOZFEIZ-UHFFFAOYSA-N | |
ethyl 5-chlorothiophene-2-carboxylate | |
521808 | |
≥98% |
70°C to 71°C (1 mmHg) | |
1.538 | |
1 g | |
5-chloro-2-thiophenecarboxylic acid ethyl ester, 5-chlorothiophene-2-carboxylic acid ethyl ester, 2-thiophenecarboxylic acid, 5-chloro-, ethyl ester, ethyl 5-chlorothiophene-2-carboxate, ethyl 5-chloro-2-thiophenecarboxylate, pubchem16133, acmc-209znt, ksc494s8t, ethyl5-chlorothiophene-2-carboxlate, ethyl 5-chloranylthiophene-2-carboxylate | |
CCOC(=O)C1=CC=C(S1)Cl | |
190.641 | |
190.65 | |
Ethyl 5-chlorothiophene-2-carboxylate |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
TSCA : No
Recommended Storage : Ambient temperatures