missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 5-chloroindole-2-carboxylate, 98%

Manufacturer:  Alfa Aesar™ L1755406

 View more versions of this product

Catalog No. AAL1755406



Specifications

Specifications

Ethyl 5-chloroindole-2-carboxylate
5g
C11H10ClNO2
170255
LWKIFKYHCJAIAB-UHFFFAOYSA-N
ethyl 5-chloro-1H-indole-2-carboxylate
78518
98%
166°C to 169°C
4792-67-0
MFCD00005610
1h-indole-2-carboxylic acid, 5-chloro-, ethyl ester
CCOC(=O)C1=CC2=C(N1)C=CC(=C2)Cl
223.656
223.66

Chemical Identifiers

4792-67-0
223.656
LWKIFKYHCJAIAB-UHFFFAOYSA-N
78518
CCOC(=O)C1=CC2=C(N1)C=CC(=C2)Cl
C11H10ClNO2
MFCD00005610
1h-indole-2-carboxylic acid, 5-chloro-, ethyl ester
ethyl 5-chloro-1H-indole-2-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents