missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 5-chloro-2-indolecarboxylate 97%, ACROS Organics™

Manufacturer:  Acros Organics 160290100

 View more versions of this product

Catalog No. AC160290100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Ethyl 5-chloro-2-indolecarboxylate
Glass bottle
166°C to 171°C
97%
97%
MFCD00005610
LWKIFKYHCJAIAB-UHFFFAOYSA-N
ethyl 5-chloro-1H-indole-2-carboxylate
78518
Powder or Crystals
Authentic
Brown or Tan to Yellow
10g
4792-67-0
C11H10ClNO2
ethyl 5-chloroindole-2-carboxylate, ethyl 5-chloro-2-indolecarboxylate, 5-chloroindole-2-carboxylic acid ethyl ester, 1h-indole-2-carboxylic acid, 5-chloro-, ethyl ester, ethyl-5-chloroindole-2-carboxylate, 5-chloro-1h-indole-2-carboxylic acid ethyl ester, pubchem7173, 5-chloroindole-2-carboxylicacidethylester, acmc-1aoif, 5-chloro-2-indolecarboxylic acid ethyl ester
CCOC(=O)C1=CC2=C(N1)C=CC(=C2)Cl
223.656
223.66
≥96.0%

Chemical Identifiers

4792-67-0
223.656
LWKIFKYHCJAIAB-UHFFFAOYSA-N
78518
CCOC(=O)C1=CC2=C(N1)C=CC(=C2)Cl
C11H10ClNO2
MFCD00005610
ethyl 5-chloroindole-2-carboxylate, ethyl 5-chloro-2-indolecarboxylate, 5-chloroindole-2-carboxylic acid ethyl ester, 1h-indole-2-carboxylic acid, 5-chloro-, ethyl ester, ethyl-5-chloroindole-2-carboxylate, 5-chloro-1h-indole-2-carboxylic acid ethyl ester, pubchem7173, 5-chloroindole-2-carboxylicacidethylester, acmc-1aoif, 5-chloro-2-indolecarboxylic acid ethyl ester
ethyl 5-chloro-1H-indole-2-carboxylate
Safety and Handling

Safety and Handling

EINECSNumber : 225-345-6

SDS Documents