missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 5-chloro-2-indolecarboxylate 97%, ACROS Organics™

Manufacturer:  Acros Organics 160290050

 View more versions of this product

Catalog No. AC160290050


 

Specifications

Specifications

Ethyl 5-chloro-2-indolecarboxylate
4792-67-0
Glass bottle
166°C to 171°C
Brown or Tan to Yellow
5g
LWKIFKYHCJAIAB-UHFFFAOYSA-N
ethyl 5-chloro-1H-indole-2-carboxylate
78518
Powder or Crystals
97%
Authentic
≥96.0%
97%
C11H10ClNO2
MFCD00005610
CCOC(=O)C1=CC2=C(N1)C=CC(=C2)Cl
223.656
223.66

Chemical Identifiers

4792-67-0
223.656
LWKIFKYHCJAIAB-UHFFFAOYSA-N
C11H10ClNO2
MFCD00005610
78518
Safety and Handling

Safety and Handling

EINECSNumber : 225-345-6

SDS Documents