missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 5-chloro-2-indolecarboxylate 97%, ACROS Organics™

$166.88 - $166.88

Chemical Identifiers

CAS 4792-67-0
Assay Percent Range 97%
Molecular Formula C11H10ClNO2
Formula Weight 223.66
MDL Number MFCD00005610
Chemical Name or Material Ethyl 5-chloro-2-indolecarboxylate, 97%
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC160290100 SDS Thermo Fisher Scientific
160290100
Glass bottle 10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC160290050 SDS Thermo Fisher Scientific
160290050
Glass bottle 5g Each for $166.88
Specifications Description & Specifications

Chemical Identifiers

4792-67-0
C11H10ClNO2
MFCD00005610
≥96.0%
97%
223.66
Ethyl 5-chloro-2-indolecarboxylate, 97%

Specifications

4792-67-0
Glass bottle
166° to 171°C
Yellow to orange to tan or light brown
10g
MFCD00005610
≥96.0%
Authentic
Powder or Crystals
97%
C11H10ClNO2
223.66
Ethyl 5-chloro-2-indolecarboxylate, 97%

Safety and Handling

EINECSNumber : 225-345-6