Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 5-bromopyrimidine-2-carboxylate, 95%

Chemical Identifiers

Chemical Name or Material Ethyl 5-bromopyrimidine-2-carboxylate
CAS 1197193-30-8
Formula Weight 231.05
MDL Number MFCD12964050
Synonym 5-Bromopyrimidine-2-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6130403

  SDS
alfa aesar™
H6130403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH61304MD

  SDS
alfa aesar™
H61304MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 231.05
Chemical Name or Material Ethyl 5-bromopyrimidine-2-carboxylate
Physical Form Crystalline
Assay 95%
Color Off-white
Quantity 1g
MDL Number MFCD12964050
CAS 1197193-30-8
Synonym 5-Bromopyrimidine-2-carboxylic acid ethyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.