missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 5-bromo-2-methylnicotinate, 97%

Manufacturer:  Alfa Aesar™ H61385MD

 View more versions of this product

Catalog No. AAH61385MD



Description

Description

Applications
Ethyl 5-bromo-2-methylnicotinate is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications

Specifications

Ethyl 5-bromo-2-methylnicotinate
250mg
C9H10BrNO2
3-pyridinecarboxylic acid, 5-bromo-2-methyl-, ethyl ester
CCOC(=O)C1=CC(=CN=C1C)Br
244.088
244.09
97%
Yellow
129477-21-0
MFCD10698571
QVKPPFGWQMCQCI-UHFFFAOYSA-N
ethyl 5-bromo-2-methylpyridine-3-carboxylate
15077476
Solid

Chemical Identifiers

129477-21-0
244.088
QVKPPFGWQMCQCI-UHFFFAOYSA-N
15077476
CCOC(=O)C1=CC(=CN=C1C)Br
C9H10BrNO2
MFCD10698571
3-pyridinecarboxylic acid, 5-bromo-2-methyl-, ethyl ester
ethyl 5-bromo-2-methylpyridine-3-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents