missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 5-Bromo-2-chlorobenzoate, TCI America™

$56.53 - $186.39

Chemical Identifiers

CAS 76008-73-6
Molecular Formula C9H8BrClO2
Molecular Weight (g/mol) 263.515
MDL Number MFCD00013635
InChI Key AMGWDYLEMSMUIO-UHFFFAOYSA-N
Synonym 5-bromo-2-chlorobenzoic acid ethyl ester, ethyl5-bromo-2-chlorobenzoate, benzoic acid, 5-bromo-2-chloro-, ethyl ester, pubchem3702, acmc-1bb9d, ethyl-5-bromo-2-chlorobenzoate, ethyl 5-bromo-2-chloro-benzoate, a-ethyl 5-bromo-2-chlorobenzoate, attercop-chm at115127, ethyl 5-bromo2-chlorobenzaldehyde
PubChem CID 144749
IUPAC Name ethyl 5-bromo-2-chlorobenzoate
SMILES CCOC(=O)C1=C(C=CC(=C1)Br)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B22555G View Documents TCI America
B22555G
5g Each for $56.53
 
B225525G View Documents TCI America
B225525G
25g Each for $186.39
 
Specifications

Chemical Identifiers

76008-73-6
263.515
AMGWDYLEMSMUIO-UHFFFAOYSA-N
144749
CCOC(=O)C1=C(C=CC(=C1)Br)Cl
C9H8BrClO2
MFCD00013635
5-bromo-2-chlorobenzoic acid ethyl ester, ethyl5-bromo-2-chlorobenzoate, benzoic acid, 5-bromo-2-chloro-, ethyl ester, pubchem3702, acmc-1bb9d, ethyl-5-bromo-2-chlorobenzoate, ethyl 5-bromo-2-chloro-benzoate, a-ethyl 5-bromo-2-chlorobenzoate, attercop-chm at115127, ethyl 5-bromo2-chlorobenzaldehyde
ethyl 5-bromo-2-chlorobenzoate

Specifications

288°C
5g
C9H8BrClO2
5-bromo-2-chlorobenzoic acid ethyl ester, ethyl5-bromo-2-chlorobenzoate, benzoic acid, 5-bromo-2-chloro-, ethyl ester, pubchem3702, acmc-1bb9d, ethyl-5-bromo-2-chlorobenzoate, ethyl 5-bromo-2-chloro-benzoate, a-ethyl 5-bromo-2-chlorobenzoate, attercop-chm at115127, ethyl 5-bromo2-chlorobenzaldehyde
CCOC(=O)C1=C(C=CC(=C1)Br)Cl
263.515
263.52
≥98.0% (GC)
Yellow
76008-73-6
MFCD00013635
AMGWDYLEMSMUIO-UHFFFAOYSA-N
ethyl 5-bromo-2-chlorobenzoate
144749
Clear Liquid at 20°C
Ethyl 5-Bromo-2-chlorobenzoate
Safety and Handling

Safety and Handling

TSCA : No