missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 5-bromo-2-chlorobenzoate, 98+%

$47.66 - $501.19

Chemical Identifiers

CAS 76008-73-6
Molecular Formula C9H8BrClO2
Molecular Weight (g/mol) 263.515
MDL Number MFCD00013635
InChI Key AMGWDYLEMSMUIO-UHFFFAOYSA-N
Synonym 5-bromo-2-chlorobenzoic acid ethyl ester, ethyl5-bromo-2-chlorobenzoate, benzoic acid, 5-bromo-2-chloro-, ethyl ester, pubchem3702, acmc-1bb9d, ethyl-5-bromo-2-chlorobenzoate, ethyl 5-bromo-2-chloro-benzoate, a-ethyl 5-bromo-2-chlorobenzoate, attercop-chm at115127, ethyl 5-bromo2-chlorobenzaldehyde
PubChem CID 144749
IUPAC Name ethyl 5-bromo-2-chlorobenzoate
SMILES CCOC(=O)C1=C(C=CC(=C1)Br)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1033406 View Documents Alfa Aesar™
A1033406
5g Each for $47.66
 
AAA1033414 View Documents Alfa Aesar™
A1033414
25g Each for $155.76
 
AAA1033422 View Documents Alfa Aesar™
A1033422
100g Each for $501.19
 
Specifications

Chemical Identifiers

76008-73-6
263.515
AMGWDYLEMSMUIO-UHFFFAOYSA-N
144749
CCOC(=O)C1=C(C=CC(=C1)Br)Cl
C9H8BrClO2
MFCD00013635
5-bromo-2-chlorobenzoic acid ethyl ester, ethyl5-bromo-2-chlorobenzoate, benzoic acid, 5-bromo-2-chloro-, ethyl ester, pubchem3702, acmc-1bb9d, ethyl-5-bromo-2-chlorobenzoate, ethyl 5-bromo-2-chloro-benzoate, a-ethyl 5-bromo-2-chlorobenzoate, attercop-chm at115127, ethyl 5-bromo2-chlorobenzaldehyde
ethyl 5-bromo-2-chlorobenzoate

Specifications

>110°C (230°F)
5g
C9H8BrClO2
6501991
AMGWDYLEMSMUIO-UHFFFAOYSA-N
ethyl 5-bromo-2-chlorobenzoate
144749
≥98%
156°C to 157°C (13mmHg)
76008-73-6
MFCD00013635
5-bromo-2-chlorobenzoic acid ethyl ester, ethyl5-bromo-2-chlorobenzoate, benzoic acid, 5-bromo-2-chloro-, ethyl ester, pubchem3702, acmc-1bb9d, ethyl-5-bromo-2-chlorobenzoate, ethyl 5-bromo-2-chloro-benzoate, a-ethyl 5-bromo-2-chlorobenzoate, attercop-chm at115127, ethyl 5-bromo2-chlorobenzaldehyde
CCOC(=O)C1=C(C=CC(=C1)Br)Cl
263.515
263.52
Ethyl 5-bromo-2-chlorobenzoate