Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 5-bromo-1,2,3-thiadiazole-4-carboxylate, 98%

$421.76 - $421.76

Chemical Identifiers

Chemical Name or Material Ethyl 5-bromo-1,2,3-thiadiazole-4-carboxylate
CAS 6439-91-4
Molecular Formula C5H5BrN2O2S
Formula Weight 237.07
MDL Number MFCD02324685
Synonym 5-Bromo-1,2,3-thiadiazole-4-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH25764MD

  SDS
alfa aesar™
H25764MD
250mg Each for $421.76
Description & Specifications

Specifications

Formula Weight 237.07
Chemical Name or Material Ethyl 5-bromo-1,2,3-thiadiazole-4-carboxylate
Assay 98%
Molecular Formula C5H5BrN2O2S
MDL Number MFCD02324685
CAS 6439-91-4
Synonym 5-Bromo-1,2,3-thiadiazole-4-carboxylic acid ethyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 76° to 78°C
Quantity 250mg

Applications
Ethyl 5-bromo-1,2,3-thiadiazole-4-carboxylate is used in the preparation of fused 1,3,6- thiadiazepines such as di[1,2,3]triazolo[1,5-a:5′,1′-d][3,1,5]benzothiadiazepines.

Solubility
Miscible with dimethyl formamide.

Notes
Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.