Please login to your online account to display your discounted pricing

Ethyl 5-amino-3-methylisothiazole-4-carboxylate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material Ethyl 5-amino-3-methylisothiazole-4-carboxylate
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H10N2O2S
Formula Weight 186.23
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

KM04205CB

  SDS
fisher scientific company
KM04205CB
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 186.23
Quantity 250mg
Assay 97%
Chemical Name or Material Ethyl 5-amino-3-methylisothiazole-4-carboxylate
Assay Percent Range 97%
Molecular Formula C7H10N2O2S
CAS 34859-65-9