Please login to your online account to display your discounted pricing

Ethyl 5-amino-3-methylisothiazole-4-carboxylate, 97%, Maybridge


Quantity 250mg

Manufacturer: fisher scientific company  KM04205CB

 View more versions of this product

Catalog No. KM04205CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 186.23
Quantity 250mg
Assay 97%
Chemical Name or Material Ethyl 5-amino-3-methylisothiazole-4-carboxylate
Molecular Formula C7H10N2O2S
Assay Percent Range 97%
CAS 34859-65-9

Specifications

Chemical Name or Material Ethyl 5-amino-3-methylisothiazole-4-carboxylate
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H10N2O2S
Formula Weight 186.23