Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 5-amino-1-phenyl-1H-pyrazole-4-carboxylate, 98%

$533.39 - $1,854.82

Chemical Identifiers

Chemical Name or Material Ethyl 5-amino-1-phenyl-1H-pyrazole-4-carboxylate
CAS 16078-71-0
Molecular Formula C12H13N3O2
Formula Weight 231.26
MDL Number MFCD00020731
Beilstein 211079
Synonym 5-Amino-4-carboethoxy-1-phenyl-1H-pyrazole; 5-Amino-1-phenyl-1H-pyrazole-4-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1329622

  SDS
alfa aesar™
A1329622
100g Each for $1,854.82

AAA1329614

  SDS
alfa aesar™
A1329614
25g Each for $533.39
Description & Specifications

Specifications

Formula Weight 231.26
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 100g
Assay 98%
CAS 16078-71-0
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym 5-Amino-4-carboethoxy-1-phenyl-1H-pyrazole; 5-Amino-1-phenyl-1H-pyrazole-4-carboxylic acid ethyl ester
Molecular Formula C12H13N3O2
RTECS Number UQ6392025
MDL Number MFCD00020731
Chemical Name or Material Ethyl 5-amino-1-phenyl-1H-pyrazole-4-carboxylate
Beilstein 211079
Melting Point 97° to 102°C
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.