missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 5-Amino-1-methylpyrazole-4-carboxylate, TCI America™

Manufacturer:  TCI America E098625G

 View more versions of this product

Catalog No. E098625G



Specifications

Specifications

Ethyl 5-Amino-1-methylpyrazole-4-carboxylate
101°C
31037-02-2
MFCD00051652
MEUSJJFWVKBUFP-UHFFFAOYSA-N
ethyl 5-amino-1-methylpyrazole-4-carboxylate
271299
Crystal-Powder at 20°C
White-Yellow
25g
C7H11N3O2
ethyl 5-amino-1-methyl-1h-pyrazole-4-carboxylate, 5-amino-1-methyl-1h-pyrazole-4-carboxylic acid ethyl ester, ethyl5-amino-1-methylpyrazole-4-carboxylate, 1h-pyrazole-4-carboxylic acid, 5-amino-1-methyl-, ethyl ester, 5-amino-1-methylpyrazole-4-carboxylic acid ethyl ester, ethyl 5-amino-1-methyl-4-pyrazolecarboxylate, ethyl 5-amino-1-methyl-pyrazole-4-carboxylate, acmc-1agpk, maybridge1_003833, ethyl-5-amino-1-methyl-1h-pyrazol-4-carboxylat
CCOC(=O)C1=C(N(N=C1)C)N
169.184
169.18
≥98.0% (GC,T)

Chemical Identifiers

31037-02-2
169.184
MEUSJJFWVKBUFP-UHFFFAOYSA-N
271299
CCOC(=O)C1=C(N(N=C1)C)N
C7H11N3O2
MFCD00051652
ethyl 5-amino-1-methyl-1h-pyrazole-4-carboxylate, 5-amino-1-methyl-1h-pyrazole-4-carboxylic acid ethyl ester, ethyl5-amino-1-methylpyrazole-4-carboxylate, 1h-pyrazole-4-carboxylic acid, 5-amino-1-methyl-, ethyl ester, 5-amino-1-methylpyrazole-4-carboxylic acid ethyl ester, ethyl 5-amino-1-methyl-4-pyrazolecarboxylate, ethyl 5-amino-1-methyl-pyrazole-4-carboxylate, acmc-1agpk, maybridge1_003833, ethyl-5-amino-1-methyl-1h-pyrazol-4-carboxylat
ethyl 5-amino-1-methylpyrazole-4-carboxylate
Safety and Handling

Safety and Handling

EINECSNumber : (5)-6164

TSCA : No

Documents