Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 5-amino-1,2,3-thiadiazole-4-carboxylate, 98+%

$331.49 - $331.49

Chemical Identifiers

Chemical Name or Material Ethyl 5-amino-1,2,3-thiadiazole-4-carboxylate
CAS 6440-02-4
Molecular Formula C5H7N3O2S
Formula Weight 173.19
MDL Number MFCD00778317
Synonym 5-Amino-1,2,3-thiadiazole-4-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH25762MD

  SDS
alfa aesar™
H25762MD
250mg Each for $331.49
Description & Specifications

Specifications

Formula Weight 173.19
Chemical Name or Material Ethyl 5-amino-1,2,3-thiadiazole-4-carboxylate
Assay ≥98%
Molecular Formula C5H7N3O2S
MDL Number MFCD00778317
CAS 6440-02-4
Synonym 5-Amino-1,2,3-thiadiazole-4-carboxylic acid ethyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 123° to 125°C
Quantity 250mg

Applications
Ethyl 5-amino-1,2,3-thiadiazole-4-carboxylate is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.