Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 5-acetyl-2-ethoxybenzoate, 98%

$197.03 - $197.03

Chemical Identifiers

Chemical Name or Material Ethyl 5-acetyl-2-ethoxybenzoate
CAS 216143-90-7
Molecular Formula C13H16O4
Formula Weight 236.27
MDL Number MFCD01075665
Synonym 4-Ethoxy-3-ethoxycarbonyl acetophenone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1475014

 
alfa aesar™
L1475014
25g Each for $197.03
Description & Specifications

Specifications

Formula Weight 236.27
Chemical Name or Material Ethyl 5-acetyl-2-ethoxybenzoate
Assay 98%
Molecular Formula C13H16O4
CAS 216143-90-7
MDL Number MFCD01075665
Synonym 4-Ethoxy-3-ethoxycarbonyl acetophenone
Melting Point 51° to 54°C
Quantity 25g