Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 5,6-dimethoxyindole-2-carboxylate, 98+%

$130.66 - $130.66

Chemical Identifiers

Chemical Name or Material Ethyl 5,6-dimethoxyindole-2-carboxylate
CAS 16382-18-6
Molecular Formula C13H15NO4
Formula Weight 249.27g/mol
MDL Number MFCD00015457
Beilstein 211948
Synonym 5,6-Dimethoxyindole-2-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0076203

 
alfa aesar™
L00762-03
1g Each for $130.66
Description & Specifications

Specifications

Melting Point 175° to 179°C
Quantity 1g
Chemical Name or Material Ethyl 5,6-dimethoxyindole-2-carboxylate
CAS 16382-18-6
Assay ≥98%
Molecular Formula C13H15NO4
Formula Weight 249.27g/mol
MDL Number MFCD00015457
Beilstein 211948
Synonym 5,6-Dimethoxyindole-2-carboxylic acid ethyl ester