missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4-(trifluoromethyl)phenylacetate, 95%

Manufacturer:  Alfa Aesar™ H6409906

 View more versions of this product

Catalog No. AAH6409906


 

Specifications

Specifications

Ethyl 4-(trifluoromethyl)phenylacetate
Colorless
95%
MFCD07779351
BDVKGYOFECBKDX-UHFFFAOYSA-N
ethyl 2-[4-(trifluoromethyl)phenyl]acetate
14134124
Solid
721-63-1
5g
C11H11F3O2
4-(Trifluoromethyl)phenylacetic acid ethyl ester
CCOC(=O)CC1=CC=C(C=C1)C(F)(F)F
232.202
232.2

Chemical Identifiers

721-63-1
232.202
BDVKGYOFECBKDX-UHFFFAOYSA-N
14134124
C11H11F3O2
MFCD07779351
4-(Trifluoromethyl)phenylacetic acid ethyl ester
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents