missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 4-(Trifluoromethyl)benzoate, TCI America™

Manufacturer:  TCI America E133025G

 View more versions of this product

Catalog No. E133025G



Specifications

Specifications

Ethyl 4-(Trifluoromethyl)benzoate
Colorless
583-02-8
MFCD00013563
ZQDADDSPMCHZPX-UHFFFAOYSA-N
ethyl 4-(trifluoromethyl)benzoate
521827
Clear Liquid at 20°C
80°C
25g
C10H9F3O2
ethyl 4-trifluoromethyl benzoate, 4-trifluoromethyl benzoic acid ethyl ester, 4-trifluoromethylbenzoic acid, ethyl ester, ethyl 4-trifluoromethyl benzoate, pubchem16913, acmc-1cs5c, ethyl-p-trifluoromethylbenzoate, ethyl-4-trifluoromethyl benzoate, ethyl-4-trifluoromethyl benzoate, 4-trifluoromethylbenzoic acid ethyl ester
CCOC(=O)C1=CC=C(C=C1)C(F)(F)F
218.175
218.18
≥98.0% (GC)

Chemical Identifiers

583-02-8
218.175
ZQDADDSPMCHZPX-UHFFFAOYSA-N
521827
CCOC(=O)C1=CC=C(C=C1)C(F)(F)F
C10H9F3O2
MFCD00013563
ethyl 4-trifluoromethyl benzoate, 4-trifluoromethyl benzoic acid ethyl ester, 4-trifluoromethylbenzoic acid, ethyl ester, ethyl 4-trifluoromethyl benzoate, pubchem16913, acmc-1cs5c, ethyl-p-trifluoromethylbenzoate, ethyl-4-trifluoromethyl benzoate, ethyl-4-trifluoromethyl benzoate, 4-trifluoromethylbenzoic acid ethyl ester
ethyl 4-(trifluoromethyl)benzoate
Safety and Handling

Safety and Handling

TSCA : No

Documents