Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4-(trifluoromethyl)benzoate, 99%

$30.32 - $30.32

Chemical Identifiers

Chemical Name or Material Ethyl 4-(trifluoromethyl)benzoate
CAS 583-02-8
Molecular Formula C10H9F3O2
Formula Weight 218.18
MDL Number MFCD00013563
Beilstein 1968247
Synonym 4-(Trifluoromethyl)benzoic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1113903

  SDS
alfa aesar™
L1113903
1g Each for $30.32
Description & Specifications

Specifications

Formula Weight 218.18
Chemical Name or Material Ethyl 4-(trifluoromethyl)benzoate
Assay 99%
Molecular Formula C10H9F3O2
MDL Number MFCD00013563
CAS 583-02-8
Synonym 4-(Trifluoromethyl)benzoic acid ethyl ester
Beilstein 1968247
Boiling Point 78° to 80°C (5mmHg)
Quantity 1g