Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4-quinazolone-2-carboxylate, 98%

$96.43 - $352.58

Chemical Identifiers

Chemical Name or Material Ethyl 4-quinazolone-2-carboxylate
CAS 29113-33-5
Molecular Formula C11H10N2O3
Formula Weight 218.21
MDL Number MFCD00051583
Beilstein 190186
Synonym 4-Quinazolone-2-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0670903

  SDS
alfa aesar™
L0670903
1g Each for $96.43

AAL0670906

  SDS
alfa aesar™
L0670906
5g Each for $352.58
Description & Specifications

Specifications

Formula Weight 218.21
Chemical Name or Material Ethyl 4-quinazolone-2-carboxylate
CAS 29113-33-5
Beilstein 190186
Assay 98%
Molecular Formula C11H10N2O3
MDL Number MFCD00051583
Synonym 4-Quinazolone-2-carboxylic acid ethyl ester
Melting Point 189° to 193°C
Quantity 1g