Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4-piperidone-3-carboxylate hydrochloride, 97%

$84.82 - $985.42

Chemical Identifiers

Chemical Name or Material Ethyl 4-piperidone-3-carboxylate hydrochloride
CAS 4644-61-5
Molecular Formula C8H13NO3·HCl
Formula Weight 207.66
MDL Number MFCD00043267
Beilstein 3711279
Synonym 3-Carboethoxy-4-piperidone hydrochloride; Ethyl 4-oxo-piperidine-3-carboxylate hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1213106

  SDS
alfa aesar™
A1213106
5g Each for $84.82

AAA1213114

  SDS
alfa aesar™
A1213114
25g Each for $280.26

AAA1213122

  SDS
alfa aesar™
A1213122
100g Each for $985.42
Description & Specifications

Specifications

Formula Weight 207.66
Chemical Name or Material Ethyl 4-piperidone-3-carboxylate hydrochloride
Assay 97%
Molecular Formula C8H13NO3·HCl
MDL Number MFCD00043267
CAS 4644-61-5
Beilstein 3711279
Synonym 3-Carboethoxy-4-piperidone hydrochloride; Ethyl 4-oxo-piperidine-3-carboxylate hydrochloride
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point ≈172°C (dec.)
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.