missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 4-Piperidinecarboxylate, TCI America™

$47.36 - $383.48

Chemical Identifiers

CAS 1126-09-6
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.213
MDL Number MFCD00006003
InChI Key RUJPPJYDHHAEEK-UHFFFAOYSA-N
Synonym ethyl 4-piperidinecarboxylate, ethyl isonipecotate, piperidine-4-carboxylic acid ethyl ester, ethylpiperidine-4-carboxylate, 4-piperidinecarboxylic acid, ethyl ester, isonipecotic acid ethyl ester, 4-piperidinecarboxylic acid ethyl ester, 4-carbethoxypiperidine, 4-carboethoxypiperidine, 4-ethoxycarbonyl piperidine
PubChem CID 70770
IUPAC Name ethyl piperidine-4-carboxylate
SMILES CCOC(=O)C1CCNCC1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I029425ML View Documents TCI America
I029425ML
25mL Each for $47.36
 
I0294500ML View Documents TCI America
I0294500ML
500mL Each for $383.48
 
I0294100ML View Documents TCI America
I0294100ML
100mL Each for $128.34
 
Specifications

Chemical Identifiers

1126-09-6
157.213
RUJPPJYDHHAEEK-UHFFFAOYSA-N
70770
CCOC(=O)C1CCNCC1
C8H15NO2
MFCD00006003
ethyl 4-piperidinecarboxylate, ethyl isonipecotate, piperidine-4-carboxylic acid ethyl ester, ethylpiperidine-4-carboxylate, 4-piperidinecarboxylic acid, ethyl ester, isonipecotic acid ethyl ester, 4-piperidinecarboxylic acid ethyl ester, 4-carbethoxypiperidine, 4-carboethoxypiperidine, 4-ethoxycarbonyl piperidine
ethyl piperidine-4-carboxylate

Specifications

105°C
25mL
C8H15NO2
ethyl 4-piperidinecarboxylate, ethyl isonipecotate, piperidine-4-carboxylic acid ethyl ester, ethylpiperidine-4-carboxylate, 4-piperidinecarboxylic acid, ethyl ester, isonipecotic acid ethyl ester, 4-piperidinecarboxylic acid ethyl ester, 4-carbethoxypiperidine, 4-carboethoxypiperidine, 4-ethoxycarbonyl piperidine
CCOC(=O)C1CCNCC1
157.213
157.21
≥98.0% (GC)
Yellow
1126-09-6
MFCD00006003
RUJPPJYDHHAEEK-UHFFFAOYSA-N
ethyl piperidine-4-carboxylate
70770
Clear Liquid at 20°C
Ethyl 4-Piperidinecarboxylate
Safety and Handling

Safety and Handling

TSCA : No