missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl 4-Phenylbutyrate 98.0+%, TCI America™
Supplier: TCI America E10275G
Specifications
Ethyl 4-Phenylbutyrate | |
Colorless | |
C12H16O2 | |
5 g | |
GGFNXKFGVQQNRV-UHFFFAOYSA-N | |
ethyl 4-phenylbutanoate | |
61452 | |
≥98.0% (GC) |
10031-93-3 | |
100°C | |
MFCD00026923 | |
4-Phenylbutyric Acid Ethyl Ester | |
CCOC(=O)CCCC1=CC=CC=C1 | |
192.258 | |
192.26 | |
Liquid |
Chemical Identifiers
10031-93-3 | |
192.258 | |
GGFNXKFGVQQNRV-UHFFFAOYSA-N | |
61452 | |
CCOC(=O)CCCC1=CC=CC=C1 |
C12H16O2 | |
MFCD00026923 | |
4-Phenylbutyric Acid Ethyl Ester | |
ethyl 4-phenylbutanoate |
Safety and Handling
EINECSNumber : (3)-1730
TSCA : No