Learn More
Ethyl 4-pentenoate, 98+%, Thermo Scientific Chemicals
$57.81 - $2140.67
Chemical Identifiers
CAS | 1968-40-7 |
---|---|
Molecular Formula | C7H12O2 |
Molecular Weight (g/mol) | 128.171 |
MDL Number | MFCD00036562 |
InChI Key | PTVSRINJXWDIKP-UHFFFAOYSA-N |
Synonym | ethyl 4-pentenoate, 4-pentenoic acid ethyl ester, 4-pentenoic acid, ethyl ester, ethyl pent-4-en-1-oate, unii-210um0f8jp, 4-ethoxycarbonylbut-1-ene, pent-4-enoic acid ethyl ester, ethyl allylacetate, ethyl 4-pentenate, ethylpent-4-enoate |
PubChem CID | 74786 |
IUPAC Name | ethyl pent-4-enoate |
SMILES | CCOC(=O)CCC=C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1242203
|
Thermo Scientific Chemicals
A1242203 |
1 g |
Each for $57.81
|
|
|||||
AAA1242206
|
Thermo Scientific Chemicals
A1242206 |
5 g |
Each for $194.40
|
|
|||||
AAA1242214
|
Thermo Scientific Chemicals
A1242214 |
25 g |
Each for $680.16
|
|
|||||
AAA1242222
|
Thermo Scientific Chemicals
A1242222 |
100 g |
Each for $2,140.67
|
|
|||||
Description
Ethyl 4-pentenoate is found in leaf oil of Prunus phaeosticta phaeosticta from Taiwan. It is also used in the preparation of 5-iodomethyl-dihydro-furan-2-one by reacting with iodine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl 4-pentenoate is found in leaf oil of Prunus phaeosticta phaeosticta from Taiwan. It is also used in the preparation of 5-iodomethyl-dihydro-furan-2-one by reacting with iodine.
Solubility
Miscible with alcohol. Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
1968-40-7 | |
128.171 | |
PTVSRINJXWDIKP-UHFFFAOYSA-N | |
74786 | |
CCOC(=O)CCC=C |
C7H12O2 | |
MFCD00036562 | |
ethyl 4-pentenoate, 4-pentenoic acid ethyl ester, 4-pentenoic acid, ethyl ester, ethyl pent-4-en-1-oate, unii-210um0f8jp, 4-ethoxycarbonylbut-1-ene, pent-4-enoic acid ethyl ester, ethyl allylacetate, ethyl 4-pentenate, ethylpent-4-enoate | |
ethyl pent-4-enoate |
Specifications
1968-40-7 | |
146°C | |
C7H12O2 | |
MFCD00036562 | |
UN3272 | |
ethyl 4-pentenoate, 4-pentenoic acid ethyl ester, 4-pentenoic acid, ethyl ester, ethyl pent-4-en-1-oate, unii-210um0f8jp, 4-ethoxycarbonylbut-1-ene, pent-4-enoic acid ethyl ester, ethyl allylacetate, ethyl 4-pentenate, ethylpent-4-enoate | |
PTVSRINJXWDIKP-UHFFFAOYSA-N | |
ethyl pent-4-enoate | |
74786 | |
≥98% |
0.9 | |
37°C (98°F) | |
1.417 | |
1 g | |
1751643 | |
Miscible with alcohol. Immiscible with water. | |
CCOC(=O)CCC=C | |
128.171 | |
128.17 | |
Ethyl 4-pentenoate |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber : 217-818-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only