Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4-oxocyclohexanecarboxylate, 97%

$90.68 - $390.97

Chemical Identifiers

Chemical Name or Material Ethyl 4-oxocyclohexanecarboxylate
CAS 17159-79-4
Molecular Formula C9H14O3
Formula Weight 170.21
MDL Number MFCD00013285
Synonym Ethyl cyclohexanone-4-carboxylate; 4-Oxocyclohexanecarboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2057203

  SDS
alfa aesar™
B2057203
1g Each for $90.68

AAB2057206

  SDS
alfa aesar™
B2057206
5g Each for $390.97
Description & Specifications

Specifications

Formula Weight 170.21
Chemical Name or Material Ethyl 4-oxocyclohexanecarboxylate
Assay 97%
Molecular Formula C9H14O3
MDL Number MFCD00013285
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
CAS 17159-79-4
Synonym Ethyl cyclohexanone-4-carboxylate; 4-Oxocyclohexanecarboxylic acid ethyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Boiling Point 150° to 152°C (40mmHg)
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.