missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 4-Oxazolecarboxylate 96.0+%, TCI America™

$53.06 - $136.44

Chemical Identifiers

CAS 23012-14-8
Molecular Formula C6H7NO3
Molecular Weight (g/mol) 141.126
MDL Number MFCD04114940
InChI Key UBESIXFCSFYQNK-UHFFFAOYSA-N
Synonym 4-Oxazolecarboxylic Acid Ethyl Ester
PubChem CID 2763217
IUPAC Name ethyl 1,3-oxazole-4-carboxylate
SMILES CCOC(=O)C1=COC=N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
O02891G
View Documents
Tci America
O02891G
1g Each for $53.06
 
O02895G
View Documents
Tci America
O02895G
5g Each for $136.44
 
Specifications

Chemical Identifiers

23012-14-8
141.126
UBESIXFCSFYQNK-UHFFFAOYSA-N
2763217
CCOC(=O)C1=COC=N1
C6H7NO3
MFCD04114940
4-Oxazolecarboxylic Acid Ethyl Ester
ethyl 1,3-oxazole-4-carboxylate

Specifications

101°C
48°C
23012-14-8
MFCD04114940
UBESIXFCSFYQNK-UHFFFAOYSA-N
ethyl 1,3-oxazole-4-carboxylate
2763217
Crystal-Powder at 20°C
Ethyl 4-Oxazolecarboxylate
White-Yellow
1g
C6H7NO3
4-Oxazolecarboxylic Acid Ethyl Ester
CCOC(=O)C1=COC=N1
141.126
141.13
≥96.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator