Please login to your online account to display your discounted pricing

Ethyl 4-nitrophenylacetate, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Ethyl 4-nitrophenylacetate
Name Note 98%
CAS 5445-26-1
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H11NO4
Linear Formula CH3CHOC(O)CHC6H4NO
Formula Weight 209.2
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC351290050

  SDS
thermo fisher scientific
351290050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 209.2
Quantity 5g
Color Light yellow
TSCA TSCA
EINECS Number 226-646-5
Melting Point 61° to 65°C
Infrared Spectrum Authentic
Linear Formula CH3CHOC(O)CHC6H4NO
Chemical Name or Material Ethyl 4-nitrophenylacetate
Name Note 98%
Assay Percent Range 98%
CAS 5445-26-1
Molecular Formula C10H11NO4