missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4-nitrophenylacetate, 98%

Manufacturer:  Alfa Aesar™ A1110106

 View more versions of this product

Catalog No. AAA1110106


 

Specifications

Specifications

Ethyl 4-nitrophenylacetate
5g
98%
MFCD00017046
4-Nitrophenylacetic acid ethyl ester
CCOC(=O)CC1=CC=C(C=C1)[N+](=O)[O-]
209.201
209.2
62°C to 66°C
5445-26-1
C10H11NO4
2053241
DWDRNKYLWMKWTH-UHFFFAOYSA-N
ethyl 2-(4-nitrophenyl)acetate
79517

Chemical Identifiers

5445-26-1
209.201
DWDRNKYLWMKWTH-UHFFFAOYSA-N
79517
C10H11NO4
MFCD00017046
4-Nitrophenylacetic acid ethyl ester
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents