missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4-nitroindole-2-carboxylate, 97%

Manufacturer:  Alfa Aesar™ H6344003

 View more versions of this product

Catalog No. AAH6344003



Specifications

Specifications

Ethyl 4-nitroindole-2-carboxylate
1g
C11H10N2O4
1h-indole-2-carboxylic acid, 4-nitro-, ethyl ester, 4-nitro-1h-indole-2-carboxylic acid ethyl ester, 4-nitro-indole-2-carboxylic acid ethyl ester, ethyl 4-nitro-2-indolecarboxylate, ethyl 4-nitroindole-2-carboxylate, ethyl-4-nitroindole-2-carboxylate
CCOC(=O)C1=CC2=C(N1)C=CC=C2[N+](=O)[O-]
234.211
234.21
97%
Yellow
4993-93-5
MFCD06795512
LXUABEANNNKOLF-UHFFFAOYSA-N
ethyl 4-nitro-1H-indole-2-carboxylate
283173
Solid

Chemical Identifiers

4993-93-5
234.211
LXUABEANNNKOLF-UHFFFAOYSA-N
283173
CCOC(=O)C1=CC2=C(N1)C=CC=C2[N+](=O)[O-]
C11H10N2O4
MFCD06795512
1h-indole-2-carboxylic acid, 4-nitro-, ethyl ester, 4-nitro-1h-indole-2-carboxylic acid ethyl ester, 4-nitro-indole-2-carboxylic acid ethyl ester, ethyl 4-nitro-2-indolecarboxylate, ethyl 4-nitroindole-2-carboxylate, ethyl-4-nitroindole-2-carboxylate
ethyl 4-nitro-1H-indole-2-carboxylate
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

Documents