Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4-nitrocinnamate, 99%

$53.19 - $607.23

Chemical Identifiers

Chemical Name or Material Ethyl 4-nitrocinnamate
CAS 953-26-4
Molecular Formula C11H11NO4
Formula Weight 221.21
MDL Number MFCD00007380
Beilstein 2052533
Synonym 4-Nitrocinnamic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1319606

  SDS
alfa aesar™
A1319606
5g Each for $53.19

AAA1319622

  SDS
alfa aesar™
A1319622
100g Each for $607.23
Description & Specifications

Specifications

Formula Weight 221.21
Chemical Name or Material Ethyl 4-nitrocinnamate
TSCA TSCA
Assay 99%
Molecular Formula C11H11NO4
MDL Number MFCD00007380
CAS 953-26-4
Beilstein 2052533
Synonym 4-Nitrocinnamic acid ethyl ester
Melting Point 138° to 140°C
Quantity 5g