Please login to your online account to display your discounted pricing

Ethyl 4-nitrobenzoylacetate 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  118760250

 View more versions of this product

Catalog No. AC118760250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Fine Crystalline Powder
Formula Weight 237.21
Quantity 25g
Color Yellow
Beilstein 10, 682
EINECS Number 212-656-7
RTECS Number AJ1073500
TSCA TSCA
Melting Point 67° to 71°C
Chemical Name or Material Ethyl 4-nitrobenzoylacetate
Name Note 98%
Assay Percent Range 98%
Molecular Formula C11H11NO5
Linear Formula O2NC6H4COCH2CO2C2H5
MDL Number MFCD00007357
CAS 838-57-3
Infrared Spectrum Authentic
Packaging Glass bottle

Specifications

Chemical Name or Material Ethyl 4-nitrobenzoylacetate
Name Note 98%
CAS 838-57-3
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C11H11NO5
Linear Formula O2NC6H4COCH2CO2C2H5
Formula Weight 237.21
MDL Number MFCD00007357