missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 4-nitrobenzoylacetate 98%, ACROS Organics™

Chemical Identifiers

CAS 838-57-3
Assay Percent Range 98%
Molecular Formula C11H11NO5
Linear Formula O2NC6H4COCH2CO2C2H5
Formula Weight 237.21
MDL Number MFCD00007357
Chemical Name or Material Ethyl 4-nitrobenzoylacetate, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC118760250 SDS Thermo Fisher Scientific
118760250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

838-57-3
C11H11NO5
237.21
Ethyl 4-nitrobenzoylacetate, 98%
98%
O2NC6H4COCH2CO2C2H5
MFCD00007357
≥97.5%

Specifications

237.21
25g
10, 682
Authentic
C11H11NO5
MFCD00007357
98%
≥97.5%
Fine Crystalline Powder
Yellow
67° to 71°C
Glass bottle
838-57-3
O2NC6H4COCH2CO2C2H5
Ethyl 4-nitrobenzoylacetate, 98%

Safety and Handling

EINECSNumber : 212-656-7

RTECSNumber : AJ1073500

TSCA : TSCA