Learn More
Ethyl 4-nitrobenzoylacetate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1760309
Description
It is employed as a reactant involved in synthesis of β-1,3-dicarbonyl aldehydes via oxidation, knoevenagel condensation using a lysine catalyst, and cycloisomerization for the synthesis of trisubstituted furans. It is also a reactant for the fluorination by HF and iodosylbenzene, intramolecular Michael addition reactions and synthesis of diols. Also applied in the synthesis of diols via reduction of aromatic and aliphatic ketos esters6 and cross-coupling reactions for stereo selective synthesis of unsymmetrical 1,4-enediones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as a reactant involved in synthesis of β-1,3-dicarbonyl aldehydes via oxidation, knoevenagel condensation using a lysine catalyst, and cycloisomerization for the synthesis of trisubstituted furans. It is also a reactant for the fluorination by HF and iodosylbenzene, intramolecular Michael addition reactions and synthesis of diols. Also applied in the synthesis of diols via reduction of aromatic and aliphatic ketos esters6 and cross-coupling reactions for stereo selective synthesis of unsymmetrical 1,4-enediones.
Notes
Store away from oxidizing agents. Keep the container tightly closed in a cool, dry place.
Specifications
Ethyl 4-nitrobenzoylacetate | |
68°C to 73°C | |
MFCD00007357 | |
1124763 | |
NGRXSVFCLHVGKU-UHFFFAOYSA-N | |
ethyl 3-(4-nitrophenyl)-3-oxopropanoate | |
13281 | |
97% |
838-57-3 | |
C11H11NO5 | |
10 g | |
ethyl 4-nitrobenzoylacetate, ethyl 3-4-nitrophenyl-3-oxopropanoate, ethyl p-nitrobenzoyl acetate, ethyl 4-nitrobenzoyl acetate, p-nitrobenzoyl acetic acid ethyl ester, ethyl p-nitrobenzoylacetate, 4-nitrobenzoylacetic acid ethyl ester, ethyl 4-nitro-beta-oxobenzenepropanoate, 4-nitrobenzoyl acetic acid ethyl ester, benzenepropanoic acid, 4-nitro-beta-oxo-, ethyl ester | |
CCOC(=O)CC(=O)C1=CC=C(C=C1)[N+](=O)[O-] | |
237.211 | |
237.21 |
Chemical Identifiers
838-57-3 | |
237.211 | |
NGRXSVFCLHVGKU-UHFFFAOYSA-N | |
13281 | |
CCOC(=O)CC(=O)C1=CC=C(C=C1)[N+](=O)[O-] |
C11H11NO5 | |
MFCD00007357 | |
ethyl 4-nitrobenzoylacetate, ethyl 3-4-nitrophenyl-3-oxopropanoate, ethyl p-nitrobenzoyl acetate, ethyl 4-nitrobenzoyl acetate, p-nitrobenzoyl acetic acid ethyl ester, ethyl p-nitrobenzoylacetate, 4-nitrobenzoylacetic acid ethyl ester, ethyl 4-nitro-beta-oxobenzenepropanoate, 4-nitrobenzoyl acetic acid ethyl ester, benzenepropanoic acid, 4-nitro-beta-oxo-, ethyl ester | |
ethyl 3-(4-nitrophenyl)-3-oxopropanoate |
Safety and Handling
EINECSNumber : 212-656-7
RTECSNumber : AJ1073500
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only