Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4-methylpyrrole-3-carboxylate, 97%

$68.50 - $885.08

Chemical Identifiers

Chemical Name or Material Ethyl 4-methylpyrrole-3-carboxylate
CAS 2199-49-7
Molecular Formula C8H11NO2
Formula Weight 153.18
MDL Number MFCD00030417
Synonym 4-Methylpyrrole-3-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2508203

  SDS
alfa aesar™
B2508203
1g Each for $68.50

AAB2508206

  SDS
alfa aesar™
B2508206
5g Each for $233.97

AAB2508214

  SDS
alfa aesar™
B2508214
25g Each for $885.08
Description & Specifications

Specifications

Formula Weight 153.18
Health Hazard 1 Warning
Assay 97%
Molecular Formula C8H11NO2
Synonym 4-Methylpyrrole-3-carboxylic acid ethyl ester
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 72° to 75°C
Chemical Name or Material Ethyl 4-methylpyrrole-3-carboxylate
MDL Number MFCD00030417
CAS 2199-49-7
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g

Applications
Ethyl 4-methylpyrrole-3-carboxylate is used as pharmaceutical intermediates

Solubility
Slightly Soluble (2.3 g/L) at 25°C

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.