missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl (4-Methylphenoxy)acetate, TCI America™

Manufacturer:  TCI America E101525G

 View more versions of this product

Catalog No. E101525G



Specifications

Specifications

Ethyl (4-Methylphenoxy)acetate
Yellow
67028-40-4
MFCD00209535
UMNOIMVMNARUSB-UHFFFAOYSA-N
ethyl 2-(4-methylphenoxy)acetate
4047281
Clear Liquid at 20°C
152°C
25g
C11H14O3
(4-Methylphenoxy)acetic Acid Ethyl Ester
CCOC(=O)COC1=CC=C(C=C1)C
194.23
194.23
≥97.0% (GC)

Chemical Identifiers

67028-40-4
194.23
UMNOIMVMNARUSB-UHFFFAOYSA-N
4047281
CCOC(=O)COC1=CC=C(C=C1)C
C11H14O3
MFCD00209535
(4-Methylphenoxy)acetic Acid Ethyl Ester
ethyl 2-(4-methylphenoxy)acetate
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0571

TSCA : Yes

Documents