Please login to your online account to display your discounted pricing

Ethyl 4-methylbenzoate 99%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC392950100

 View more versions of this product

Catalog No. AC392950100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 164.2
Physical Form Liquid
Quantity 10g
Color Clear colorless to pale yellow
Density 1.2500g/mL
Packaging Glass bottle
Assay Percent Range 99%
Chemical Name or Material Ethyl 4-methylbenzoate
MDL Number MFCD00009117
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Refractive Index 1.5070 to 1.5090 (20°C, 589nm)
Infrared Spectrum Authentic
Molecular Formula C10H12O2
CAS 94-08-6

Specifications

Chemical Name or Material Ethyl 4-methylbenzoate
CAS 94-08-6
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H12O2
Formula Weight 164.2
MDL Number MFCD00009117
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents