Please login to your online account to display your discounted pricing

Ethyl 4-methylbenzoate 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Ethyl 4-methylbenzoate
CAS 94-08-6
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H12O2
Formula Weight 164.2
MDL Number MFCD00009117
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC392950100

  SDS
thermo fisher scientific
AC392950100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC392950500

  SDS
thermo fisher scientific
AC392950500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 164.2
Quantity 10g
Color Clear colorless to pale yellow
Density 1.2500g/mL
Chemical Name or Material Ethyl 4-methylbenzoate
Packaging Glass bottle
Refractive Index 1.5070 to 1.5090 (20°C, 589nm)
Assay Percent Range 99%
Molecular Formula C10H12O2
MDL Number MFCD00009117
CAS 94-08-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.