missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 4-Methyl-2-pyrrolecarboxylate, TCI America™

$261.25 - $924.33

Chemical Identifiers

CAS 40611-85-6
Molecular Formula C8H11NO2
Molecular Weight (g/mol) 153.181
MDL Number MFCD00191668
InChI Key RWFKYBVNHRKZSN-UHFFFAOYSA-N
Synonym 4-Methyl-2-pyrrolecarboxylic Acid Ethyl Ester
PubChem CID 643361
IUPAC Name ethyl 4-methyl-1H-pyrrole-2-carboxylate
SMILES CCOC(=O)C1=CC(=CN1)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M11451G View Documents TCI America
M11451G
1g Each for $261.25
 
M11455G View Documents TCI America
M11455G
5g Each for $924.33
 
Specifications

Chemical Identifiers

40611-85-6
153.181
RWFKYBVNHRKZSN-UHFFFAOYSA-N
643361
CCOC(=O)C1=CC(=CN1)C
C8H11NO2
MFCD00191668
4-Methyl-2-pyrrolecarboxylic Acid Ethyl Ester
ethyl 4-methyl-1H-pyrrole-2-carboxylate

Specifications

104°C
43°C
40611-85-6
MFCD00191668
RWFKYBVNHRKZSN-UHFFFAOYSA-N
ethyl 4-methyl-1H-pyrrole-2-carboxylate
643361
Crystal-Powder at 20°C
Ethyl 4-Methyl-2-pyrrolecarboxylate
White
1g
C8H11NO2
4-Methyl-2-pyrrolecarboxylic Acid Ethyl Ester
CCOC(=O)C1=CC(=CN1)C
153.181
153.18
≥97.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No