missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4-methyl-2-(3-pyridyl)thiazole-5-carboxylate, 97%

Manufacturer:  Alfa Aesar™ H5494206

 View more versions of this product

Catalog No. AAH5494206



Description

Description

Applications
Ethyl 4-methyl-2-(3-pyridyl)thiazole-5-carboxylate is used as a catalytic agent and petrochemical additive.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

Ethyl 4-methyl-2-(3-pyridyl)thiazole-5-carboxylate
39091-00-4
MFCD01567252
PNJYSPUXYJWHJJ-UHFFFAOYSA-N
ethyl 4-methyl-2-pyridin-3-yl-1,3-thiazole-5-carboxylate
619088
97%
5g
C12H12N2O2S
4-methyl-2-pyridin-3-yl-thiazole-5-carboxylic acid ethyl ester
CCOC(=O)C1=C(N=C(S1)C2=CN=CC=C2)C
248.3
248.3

Chemical Identifiers

39091-00-4
248.3
PNJYSPUXYJWHJJ-UHFFFAOYSA-N
619088
CCOC(=O)C1=C(N=C(S1)C2=CN=CC=C2)C
C12H12N2O2S
MFCD01567252
4-methyl-2-pyridin-3-yl-thiazole-5-carboxylic acid ethyl ester
ethyl 4-methyl-2-pyridin-3-yl-1,3-thiazole-5-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents