missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4-methyl-2-(2-pyridyl)thiazole-5-carboxylate, 97%

Manufacturer:  Alfa Aesar™ H54842MD

 View more versions of this product

Catalog No. AAH54842MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Ethyl 4-methyl-2-(2-pyridyl)thiazole-5-carboxylate
383146-88-1
MFCD01936157
UGCVLLXXBVTJPK-UHFFFAOYSA-N
ethyl 4-methyl-2-pyridin-2-yl-1,3-thiazole-5-carboxylate
2763623
97%
250mg
C12H12N2O2S
ethyl 4-methyl-2-2-pyridinyl-1,3-thiazole-5-carboxylate
CCOC(=O)C1=C(N=C(S1)C2=CC=CC=N2)C
248.3
248.3

Chemical Identifiers

383146-88-1
248.3
UGCVLLXXBVTJPK-UHFFFAOYSA-N
2763623
CCOC(=O)C1=C(N=C(S1)C2=CC=CC=N2)C
C12H12N2O2S
MFCD01936157
ethyl 4-methyl-2-2-pyridinyl-1,3-thiazole-5-carboxylate
ethyl 4-methyl-2-pyridin-2-yl-1,3-thiazole-5-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents