missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4-methyl-2-(2-pyridyl)thiazole-5-carboxylate, 97%

Manufacturer:  Alfa Aesar™ H5484206

 View more versions of this product

Catalog No. AAH5484206

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Ethyl 4-methyl-2-(2-pyridyl)thiazole-5-carboxylate
383146-88-1
MFCD01936157
UGCVLLXXBVTJPK-UHFFFAOYSA-N
ethyl 4-methyl-2-pyridin-2-yl-1,3-thiazole-5-carboxylate
2763623
97%
5g
C12H12N2O2S
ethyl 4-methyl-2-2-pyridinyl-1,3-thiazole-5-carboxylate
CCOC(=O)C1=C(N=C(S1)C2=CC=CC=N2)C
248.3
248.3

Chemical Identifiers

383146-88-1
248.3
UGCVLLXXBVTJPK-UHFFFAOYSA-N
2763623
CCOC(=O)C1=C(N=C(S1)C2=CC=CC=N2)C
C12H12N2O2S
MFCD01936157
ethyl 4-methyl-2-2-pyridinyl-1,3-thiazole-5-carboxylate
ethyl 4-methyl-2-pyridin-2-yl-1,3-thiazole-5-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents