missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 4-Methoxycinnamate, TCI America™

$129.86 - $371.25

Chemical Identifiers

CAS 24393-56-4
Molecular Formula C12H14O3
Molecular Weight (g/mol) 206.241
MDL Number MFCD00026906
InChI Key DHNGCHLFKUPGPX-RMKNXTFCSA-N
Synonym 4-Methoxycinnamic Acid Ethyl Ester, Ethyl 3-(4-Methoxyphenyl)acrylate, 3-(4-Methoxyphenyl)acrylic Acid Ethyl Ester
PubChem CID 5281783
ChEBI CHEBI:29015
IUPAC Name ethyl (E)-3-(4-methoxyphenyl)prop-2-enoate
SMILES CCOC(=O)C=CC1=CC=C(C=C1)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M12045G View Documents TCI America
M12045G
5g Each for $129.86
 
M120425G View Documents TCI America
M120425G
25g Each for $371.25
 
Specifications

Chemical Identifiers

24393-56-4
206.241
DHNGCHLFKUPGPX-RMKNXTFCSA-N
5281783
ethyl (E)-3-(4-methoxyphenyl)prop-2-enoate
C12H14O3
MFCD00026906
4-Methoxycinnamic Acid Ethyl Ester, Ethyl 3-(4-Methoxyphenyl)acrylate, 3-(4-Methoxyphenyl)acrylic Acid Ethyl Ester
CHEBI:29015
CCOC(=O)C=CC1=CC=C(C=C1)OC

Specifications

187°C
49°C
24393-56-4
MFCD00026906
DHNGCHLFKUPGPX-RMKNXTFCSA-N
ethyl (E)-3-(4-methoxyphenyl)prop-2-enoate
5281783
206.24
≥98.0% (GC)
White
5g
C12H14O3
4-Methoxycinnamic Acid Ethyl Ester, Ethyl 3-(4-Methoxyphenyl)acrylate, 3-(4-Methoxyphenyl)acrylic Acid Ethyl Ester
CCOC(=O)C=CC1=CC=C(C=C1)OC
206.241
CHEBI:29015
Crystal-Powder at 20°C
Ethyl 4-Methoxycinnamate
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1706

TSCA : No